siRNA Details
| Gene Symbol: LINC00917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024406.1 | 7 | 1861 | |||
| GenBank mRNA | AK093244.1 | - | 7 | 1861 | ||
| BC043250.1 | - | 6 | 1918 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL137562.1 | - | 5 | 769 | Chr. 16 - 86331844 - 86337556 [-] on Build GRCh38 | |

LINC00917
Genomic Map