siRNA Details
| Gene Symbol: LOC101926975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110558.1 | 1 | 71 | |||
| GenBank mRNA | AL833138.1 | - | 1 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926975| Gene Symbol: LOC101926975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110558.1 | 1 | 71 | |||
| GenBank mRNA | AL833138.1 | - | 1 | 72 | ||