siRNA Details
Gene Symbol: PP14571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024014.1 | 2 | 1541 | |||
GenBank mRNA | AF070536.1 | - | 1 | 951 | ||
AK092442.1 | - | 2 | 1541 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PP14571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024014.1 | 2 | 1541 | |||
GenBank mRNA | AF070536.1 | - | 1 | 951 | ||
AK092442.1 | - | 2 | 1541 |