siRNA Details
| Gene Symbol: MYCNUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125783.1 | 3 | 914 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYCNUT| Gene Symbol: MYCNUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125783.1 | 3 | 914 | |||