siRNA Details
Gene Symbol: LOC100507642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108064.1 | 3 | 822 | |||
NR_108065.1 | 2 | 715 | ||||
GenBank mRNA | AK024243.1 | - | 2 | 3251 | ||
AK310146.1 | - | 2 | 689 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI770426.1 | - | 2 | 654 | Chr. 7 - 149786 - 153815 [+] on Build GRCh38 |
