siRNA Details
Gene Symbol: LINC00114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027065.1 | 7 | 711 | |||
NR_027066.1 | 6 | 748 | ||||
NR_027067.1 | 4 | 432 | ||||
GenBank mRNA | AY204744.1 | - | 5 | 536 | ||
AY204745.1 | - | 6 | 748 | |||
AY204746.1 | - | 7 | 711 | |||
AY204747.1 | - | 428 | ||||
AY204750.1 | - | 4 | 1645 |
