siRNA Details
Gene Symbol: RUSC1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039517.1 | NP_001034606.1 | 2 | 1226 | ||
GenBank mRNA | BC062571.1 | - | 2 | 1255 | 5183308 | |
BC080538.1 | - | 2 | 1226 | 5018492 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY316999.1 | - | 1 | 159 | Chr. 1 - 155319653 - 155320665 [-] on Build GRCh38 |
