siRNA Details
| Gene Symbol: LOC105371907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136503.1 | 1, 2 | 296 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371907| Gene Symbol: LOC105371907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136503.1 | 1, 2 | 296 | |||