siRNA Details
Gene Symbol: LOC100507557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038244.1 | 4 | 800 | |||
NR_038245.1 | 3 | 694 | ||||
NR_038246.1 | 3 | 226 | ||||
GenBank mRNA | AK124757.1 | - | 3 | 223 | ||
AK311235.1 | - | 3 | 658 | |||
BC043592.1 | - | 4 | 434 | |||
DB465589.1 | - | 3 | 190 |
