siRNA Details
| Gene Symbol: LOC100507557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038244.1 | 4 | 3352 | |||
| NR_038245.1 | 3 | 3246 | ||||
| NR_038246.1 | 3 | 2778 | ||||
| GenBank mRNA | AK124757.1 | - | 3 | 2775 | ||
| AK311235.1 | - | 3 | 3210 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507557| Gene Symbol: LOC100507557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038244.1 | 4 | 3352 | |||
| NR_038245.1 | 3 | 3246 | ||||
| NR_038246.1 | 3 | 2778 | ||||
| GenBank mRNA | AK124757.1 | - | 3 | 2775 | ||
| AK311235.1 | - | 3 | 3210 | |||