siRNA Details
| Gene Symbol: LOC643355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270945.1 | NP_001257874.1 | 2 | 1124 | ||
| GenBank mRNA | BC034233.1 | - | 2 | 1156 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC643355| Gene Symbol: LOC643355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270945.1 | NP_001257874.1 | 2 | 1124 | ||
| GenBank mRNA | BC034233.1 | - | 2 | 1156 | ||