siRNA Details
| Gene Symbol: LOC100507461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134937.1 | 2 | 228 | |||
| GenBank mRNA | AA393442.1 | - | 2 | 230 | ||
| BX096822.1 | - | 2 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507461| Gene Symbol: LOC100507461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134937.1 | 2 | 228 | |||
| GenBank mRNA | AA393442.1 | - | 2 | 230 | ||
| BX096822.1 | - | 2 | 183 | |||