siRNA Details
| Gene Symbol: C21orf91-OT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038870.1 | 3 | 244 | |||
| NR_038871.1 | 4 | 309 | ||||
| GenBank mRNA | AY063452.1 | - | 4 | 309 | ||
| AY063453.1 | - | 3 | 244 | |||
| BX099338.1 | - | 3 | 245 | |||
| CR745563.1 | - | 3 | 243 | |||

C21orf91-OT1
Genomic Map