siRNA Details
| Gene Symbol: BANCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047671.2 | 2 | 232 | |||
| GenBank mRNA | N98863.1 | - | 2 | 140 | ||
| W58600.1 | - | 2 | 74 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BANCR| Gene Symbol: BANCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047671.2 | 2 | 232 | |||
| GenBank mRNA | N98863.1 | - | 2 | 140 | ||
| W58600.1 | - | 2 | 74 | |||