siRNA Details
| Gene Symbol: LINC01606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038235.2 | 10 | 1939 | |||
| GenBank mRNA | CR749704.1 | - | 8 | 1736 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01606| Gene Symbol: LINC01606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038235.2 | 10 | 1939 | |||
| GenBank mRNA | CR749704.1 | - | 8 | 1736 | ||