siRNA Details
Gene Symbol: TRAM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103446.1 | 3 | 1884 | |||
NR_103447.1 | 4 | 1390 | ||||
GenBank mRNA | AK095117.1 | - | 3 | 1779 | ||
CX787946.1 | - | 2 | 524 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TRAM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103446.1 | 3 | 1884 | |||
NR_103447.1 | 4 | 1390 | ||||
GenBank mRNA | AK095117.1 | - | 3 | 1779 | ||
CX787946.1 | - | 2 | 524 |