siRNA Details
| Gene Symbol: LOC100132147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135764.1 | 2 | 1402 | |||
| GenBank mRNA | BC036435.1 | - | 2 | 1404 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100132147| Gene Symbol: LOC100132147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135764.1 | 2 | 1402 | |||
| GenBank mRNA | BC036435.1 | - | 2 | 1404 | ||