siRNA Details
| Gene Symbol: LINC01339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120601.1 | 1, 2 | 175 | |||
| GenBank mRNA | AW514364.1 | - | 1, 2 | 175 | ||
| BX105792.1 | - | 1, 2 | 150 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01339| Gene Symbol: LINC01339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120601.1 | 1, 2 | 175 | |||
| GenBank mRNA | AW514364.1 | - | 1, 2 | 175 | ||
| BX105792.1 | - | 1, 2 | 150 | |||