siRNA Details
| Gene Symbol: LINC01478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110792.1 | 4, 5 | 897 | |||
| GenBank mRNA | BC051727.1 | - | 4, 5 | 897 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01478| Gene Symbol: LINC01478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110792.1 | 4, 5 | 897 | |||
| GenBank mRNA | BC051727.1 | - | 4, 5 | 897 | ||