siRNA Details
| Gene Symbol: TEX26-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038287.1 | 1 | 206 | |||
| NR_038288.1 | 1 | 206 | ||||
| GenBank mRNA | AK093165.1 | - | 1 | 206 | ||
| BC042736.1 | - | 1 | 181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX26-AS1| Gene Symbol: TEX26-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038287.1 | 1 | 206 | |||
| NR_038288.1 | 1 | 206 | ||||
| GenBank mRNA | AK093165.1 | - | 1 | 206 | ||
| BC042736.1 | - | 1 | 181 | |||