siRNA Details
| Gene Symbol: LOC101929172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104677.1 | 3 | 1548 | |||
| GenBank mRNA | AK057515.1 | - | 3 | 1548 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929172| Gene Symbol: LOC101929172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104677.1 | 3 | 1548 | |||
| GenBank mRNA | AK057515.1 | - | 3 | 1548 | ||