siRNA Details
| Gene Symbol: DPH6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038251.1 | 3 | 1203 | |||
| GenBank mRNA | BC025664.1 | - | 3 | 1203 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPH6-AS1| Gene Symbol: DPH6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038251.1 | 3 | 1203 | |||
| GenBank mRNA | BC025664.1 | - | 3 | 1203 | ||