siRNA Details
Gene Symbol: CH17-340M24.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103768.1 | 2 | 416 | |||
GenBank mRNA | BC009467.1 | - | 2 | 386 | ||
BQ889876.1 | - | 2 | 416 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CH17-340M24.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103768.1 | 2 | 416 | |||
GenBank mRNA | BC009467.1 | - | 2 | 386 | ||
BQ889876.1 | - | 2 | 416 |