siRNA Details
| Gene Symbol: CH17-340M24.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103768.1 | 1 | 34 | |||
| GenBank mRNA | BF674246.1 | - | 1 | 14 | ||
| BQ889876.1 | - | 1 | 34 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CH17-340M24.3| Gene Symbol: CH17-340M24.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103768.1 | 1 | 34 | |||
| GenBank mRNA | BF674246.1 | - | 1 | 14 | ||
| BQ889876.1 | - | 1 | 34 | |||