siRNA Details
| Gene Symbol: CSNK1G2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033400.1 | 3 | 1230 | |||
| GenBank mRNA | BC029585.1 | - | 4 | 1175 | 5271543 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSNK1G2-AS1| Gene Symbol: CSNK1G2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033400.1 | 3 | 1230 | |||
| GenBank mRNA | BC029585.1 | - | 4 | 1175 | 5271543 | |