siRNA Details
| Gene Symbol: CARD8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040599.1 | 2 | 713 | |||
| GenBank mRNA | AK125548.1 | - | 2 | 713 | ||
| BC008448.1 | - | 2 | 692 | |||
| BC060888.1 | - | 2 | 699 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CARD8-AS1| Gene Symbol: CARD8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040599.1 | 2 | 713 | |||
| GenBank mRNA | AK125548.1 | - | 2 | 713 | ||
| BC008448.1 | - | 2 | 692 | |||
| BC060888.1 | - | 2 | 699 | |||