siRNA Details
| Gene Symbol: LOC101929125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109859.1 | 4 | 679 | |||
| GenBank mRNA | BC042140.1 | - | 4 | 679 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ051453.1 | - | 4 | 677 | Chr. 20 - 3808496 - 3812422 [+] on Build GRCh38 | |

LOC101929125
Genomic Map