siRNA Details
| Gene Symbol: LOC101929631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110794.1 | 1 | 935 | |||
| GenBank mRNA | BC039113.1 | - | 1 | 938 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929631| Gene Symbol: LOC101929631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110794.1 | 1 | 935 | |||
| GenBank mRNA | BC039113.1 | - | 1 | 938 | ||