siRNA Details
| Gene Symbol: C1orf195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278501.1 | NP_001265430.1 | 1 | 83 | ||
| NM_001278502.1 | NP_001265431.1 | 1 | 83 | |||
| XM_017002222.1 | XP_016857711.1 | 1 | 675 | |||
| GenBank mRNA | BX095243.1 | - | 1 | 83 | ||

C1orf195
Genomic Map