siRNA Details
| Gene Symbol: C1orf195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278502.1 | NP_001265431.1 | 2 | 194 | ||
| XM_017002222.1 | XP_016857711.1 | 2 | 786 | |||
| GenBank mRNA | BX095243.1 | - | 2 | 194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf195| Gene Symbol: C1orf195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278502.1 | NP_001265431.1 | 2 | 194 | ||
| XM_017002222.1 | XP_016857711.1 | 2 | 786 | |||
| GenBank mRNA | BX095243.1 | - | 2 | 194 | ||