siRNA Details
Gene Symbol: LINC01534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110707.1 | 2 | 276 | |||
NR_110708.1 | 2 | 276 | ||||
GenBank mRNA | BM701845.1 | - | 2 | 277 | ||
BQ049787.1 | - | 2 | 237 | |||
DA064533.1 | - | 2 | 261 | |||
DA195154.1 | - | 2 | 261 |
