siRNA Details
| Gene Symbol: LINC01180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110661.1 | 1 | 1419 | |||
| GenBank mRNA | BC043289.1 | - | 1 | 1423 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01180| Gene Symbol: LINC01180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110661.1 | 1 | 1419 | |||
| GenBank mRNA | BC043289.1 | - | 1 | 1423 | ||