siRNA Details
| Gene Symbol: LINC01473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110218.1 | 6 | 358 | |||
| GenBank mRNA | BM725722.1 | - | 5 | 412 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01473| Gene Symbol: LINC01473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110218.1 | 6 | 358 | |||
| GenBank mRNA | BM725722.1 | - | 5 | 412 | ||