siRNA Details
Gene Symbol: LOC101928710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110848.1 | 5 | 2190 | |||
NR_110849.1 | 5 | 881 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG203385.1 | - | 2 | 299 | Chr. 17 - 78617388 - 78624318 [-] on Build GRCh38 |
