siRNA Details
| Gene Symbol: LOC101928710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110848.1 | 4 | 507 | |||
| NR_110849.1 | 4 | 616 | ||||
| GenBank mRNA | BG613706.1 | - | 4 | 617 | ||
| BX362577.2 | - | 4 | 501 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG203385.1 | - | 1 | 34 | Chr. 17 - 78617388 - 78624318 [-] on Build GRCh38 | |

LOC101928710
Genomic Map