siRNA Details
| Gene Symbol: POT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125718.1 | 2 | 224 | |||
| NR_125719.1 | 2 | 224 | ||||
| GenBank mRNA | AK090675.1 | - | 2 | 199 | ||
| BC142949.1 | - | 2 | 182 | |||
| BX648695.1 | - | 2 | 183 | |||
| DA063136.1 | - | 2 | 224 | |||

POT1-AS1
Genomic Map