siRNA Details
| Gene Symbol: ROPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006714503.2 | XP_006714566.1 | 6 | 976 | ||
| XM_006714504.2 | XP_006714567.1 | 7 | 878 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ROPN1L| Gene Symbol: ROPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006714503.2 | XP_006714566.1 | 6 | 976 | ||
| XM_006714504.2 | XP_006714567.1 | 7 | 878 | |||