siRNA Details
| Gene Symbol: TRHDE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026837.1 | 2 | 2943 | |||
| GenBank mRNA | AK095156.1 | - | 2 | 1960 | ||
| CR936828.1 | - | 1 | 3726 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRHDE-AS1| Gene Symbol: TRHDE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026837.1 | 2 | 2943 | |||
| GenBank mRNA | AK095156.1 | - | 2 | 1960 | ||
| CR936828.1 | - | 1 | 3726 | |||