siRNA Details
| Gene Symbol: LOC284788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027089.2 | 1 | 369 | |||
| NR_027090.1 | 1 | 369 | ||||
| GenBank mRNA | BC033532.1 | - | 1 | 369 | ||
| BC144090.1 | - | 1 | 241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284788| Gene Symbol: LOC284788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027089.2 | 1 | 369 | |||
| NR_027090.1 | 1 | 369 | ||||
| GenBank mRNA | BC033532.1 | - | 1 | 369 | ||
| BC144090.1 | - | 1 | 241 | |||