siRNA Details
Gene Symbol: LOC101926905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105056.2 | 1 | 103 | |||
GenBank mRNA | HY010779.1 | - | 1 | 96 | ||
HY018974.1 | - | 1 | 105 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101926905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105056.2 | 1 | 103 | |||
GenBank mRNA | HY010779.1 | - | 1 | 96 | ||
HY018974.1 | - | 1 | 105 |