siRNA Details
| Gene Symbol: LOC101926905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105056.2 | 3 | 468 | |||
| GenBank mRNA | BG193185.1 | - | 2 | 235 | ||
| HY003594.1 | - | 4 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926905| Gene Symbol: LOC101926905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105056.2 | 3 | 468 | |||
| GenBank mRNA | BG193185.1 | - | 2 | 235 | ||
| HY003594.1 | - | 4 | 406 | |||