siRNA Details
| Gene Symbol: LOC401463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015374.1 | 2 | 2351 | |||
| GenBank mRNA | AK097150.1 | - | 2 | 2141 | ||
| BC022555.1 | - | 2 | 2351 | |||
| BC050012.1 | - | 2 | 2351 | |||
| BC068582.1 | - | 2 | 2351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC401463| Gene Symbol: LOC401463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015374.1 | 2 | 2351 | |||
| GenBank mRNA | AK097150.1 | - | 2 | 2141 | ||
| BC022555.1 | - | 2 | 2351 | |||
| BC050012.1 | - | 2 | 2351 | |||
| BC068582.1 | - | 2 | 2351 | |||