siRNA Details
Gene Symbol: LINC00943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038256.1 | 4 | 1306 | |||
GenBank mRNA | AK091521.1 | - | 3 | 1896 | ||
AY070435.1 | - | 1672 | ||||
BC039096.1 | - | 4 | 1288 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB516401.1 | - | 2 | 418 | Chr. 12 - 126744798 - 126745336 [+] on Build GRCh38 |
