siRNA Details
| Gene Symbol: THCAT158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110880.1 | 3 | 622 | |||
| NR_110881.1 | 3 | 486 | ||||
| GenBank mRNA | AA430297.1 | - | 3 | 486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

THCAT158| Gene Symbol: THCAT158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110880.1 | 3 | 622 | |||
| NR_110881.1 | 3 | 486 | ||||
| GenBank mRNA | AA430297.1 | - | 3 | 486 | ||