siRNA Details
Gene Symbol: LOC101929159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135550.1 | 2 | 2296 | |||
GenBank mRNA | BC044922.2 | - | 2 | 2302 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135550.1 | 2 | 2296 | |||
GenBank mRNA | BC044922.2 | - | 2 | 2302 |