siRNA Details
Gene Symbol: LOC101929698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110619.1 | 1 | 93 | |||
GenBank mRNA | AK097865.1 | - | 1 | 93 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR610740.1 | - | 1 | 45 | Chr. 20 - 32510007 - 32511863 [+] on Build GRCh38 |
