siRNA Details
| Gene Symbol: C1QTNF1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040018.1 | 3 | 814 | |||
| NR_040019.1 | 3 | 767 | ||||
| GenBank mRNA | BC039683.1 | - | 3 | 713 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1QTNF1-AS1| Gene Symbol: C1QTNF1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040018.1 | 3 | 814 | |||
| NR_040019.1 | 3 | 767 | ||||
| GenBank mRNA | BC039683.1 | - | 3 | 713 | ||