siRNA Details
| Gene Symbol: LINC01607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125410.1 | 1, 2 | 84 | |||
| GenBank mRNA | AI214797.1 | - | 1, 2 | 84 | ||
| AI801879.1 | - | 1, 2 | 85 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01607| Gene Symbol: LINC01607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125410.1 | 1, 2 | 84 | |||
| GenBank mRNA | AI214797.1 | - | 1, 2 | 84 | ||
| AI801879.1 | - | 1, 2 | 85 | |||