siRNA Details
| Gene Symbol: LOC284950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038888.1 | 3 | 677 | |||
| GenBank mRNA | AK095038.1 | - | 3 | 678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284950| Gene Symbol: LOC284950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038888.1 | 3 | 677 | |||
| GenBank mRNA | AK095038.1 | - | 3 | 678 | ||