siRNA Details
| Gene Symbol: LOC105376736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135061.1 | 2 | 478 | |||
| NR_135062.1 | 2 | 382 | ||||
| NR_135063.1 | 2 | 308 | ||||
| GenBank mRNA | AI342698.1 | - | 2 | 382 | ||
| BG184665.1 | - | 1 | 248 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376736| Gene Symbol: LOC105376736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135061.1 | 2 | 478 | |||
| NR_135062.1 | 2 | 382 | ||||
| NR_135063.1 | 2 | 308 | ||||
| GenBank mRNA | AI342698.1 | - | 2 | 382 | ||
| BG184665.1 | - | 1 | 248 | |||